Thermo Scientific Chemicals

Statsafe^t 6000, Thermo Scientific Chemicals

Catalog number: H55787.22
100 g, Each
Thermo Scientific Chemicals

Statsafe^t 6000, Thermo Scientific Chemicals

Catalog number: H55787.22
100 g, Each
Quantity
Catalog number: H55787.22
also known as H55787-22
Price (USD)
Quantity
-
Have Questions?
Comment
Product not available for purchase

Description

Static dissipater used in many chemical processing operations to reduce the risk of static build up and consequent damage and loss. Especially recommended for use in the manufacturing of polyolefins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Static dissipater used in many chemical processing operations to reduce the risk of static build up and consequent damage and loss. The Protein Crystallography Extension Kit substantially increases the screening range of the Basic Crystallography Kit by providing a broader range of organic solvents, PEGs, and additives (divalent ions and stabilizers).

Solubility
Not miscible with water.

Notes
Suitable additive to increase hydrocarbon conductivity and to reduce electrostatic hazards in a variety of applications.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text