Thermo Scientific Chemicals

2,2'-Diaminodiphenyl disulfide, 97%, Thermo Scientific Chemicals

Catalog number: H55807.06
5 g, Each
Thermo Scientific Chemicals

2,2'-Diaminodiphenyl disulfide, 97%, Thermo Scientific Chemicals

Catalog number: H55807.06
5 g, Each
Quantity
Catalog number: H55807.06
also known as H55807-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1141-88-4
IUPAC Name
2-[(2-aminophenyl)disulfanyl]aniline
Molecular Formula
C12H12N2S2
InChI Key
YYYOQURZQWIILK-UHFFFAOYSA-N
SMILES
NC1=CC=CC=C1SSC1=CC=CC=C1N
Assay (HPLC)
>96.0%

Description

2,2'-Diaminodiphenyl disulfide, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as a primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′-Diaminodiphenyl disulfide, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as a primary and secondary intermediate.

Solubility
Soluble in hot methanol- almost transparent.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text