N-Allylaniline, 95%, Thermo Scientific Chemicals
N-Allylaniline, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Allylaniline, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number H55821.14
also known as H55821-14
Price (USD)/ Each
121.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
121.00
Add to cart
N-Allylaniline, 95%, Thermo Scientific Chemicals
Catalog numberH55821.14
Price (USD)/ Each
121.00
-
Add to cart
Chemical Identifiers
CAS589-09-3
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)>94.0%
Refractive Index1.5620-1.5660 @ 20?C
Appearance (Color)Clear colorless to orange to brown
It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Keep away from oxidizing agents and strong acids.
RUO – Research Use Only

General References:

  1. M. Aresta.; M. De Fazio. Metal complexes with allylanilines ☆: IV. Synthesis and reactivity of rhodium(I) complexes with 2-allylaniline and N-allylaniline. J. Organomet. Chem. 1980, 186 (1),109-120.
  2. Roy Odle.; Burke Blevins.; Matt Ratcliff.; Louis S. Hegedus. Conversion of 2-halo-N-allylanilines to indoles via palladium(0) oxidative addition-insertion reactions.J. Org. Chem. 1980, 45 (13),2709-2710.