Thermo Scientific Chemicals

4-Methoxybenzenediazonium tetrafluoroborate, 98%, Thermo Scientific Chemicals

Catalog number: H55827.03
1 g, Each
Thermo Scientific Chemicals

4-Methoxybenzenediazonium tetrafluoroborate, 98%, Thermo Scientific Chemicals

Catalog number: H55827.03
1 g, Each
Quantity
Catalog number: H55827.03
also known as H55827-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
459-64-3
IUPAC Name
boron(3+) 4-methoxybenzene-1-diazonium tetrafluoride
Molecular Formula
C7H7BF4N2O
InChI Key
CACFTKIREZJSIG-UHFFFAOYSA-J
SMILES
[B+3].[F-].[F-].[F-].[F-].COC1=CC=C(C=C1)[N+]#N
Form
Powder
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Appearance (Color)
White to cream to pale brown

Description

4-Methoxybenzenediazonium tetrafluoroborate has been used in the preparation of azo coupled cyclic β-enaminones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxybenzenediazonium tetrafluoroborate has been used in the preparation of azo coupled cyclic β-enaminones.

Solubility
Decompose in water

Notes
Store at -20°C. Light sensitive.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text