Thermo Scientific Chemicals

4-Fluorobenzenesulfonamide, 98+%, Thermo Scientific Chemicals

Catalog number: H55834.MD
250 mg, Each
Thermo Scientific Chemicals

4-Fluorobenzenesulfonamide, 98+%, Thermo Scientific Chemicals

Catalog number: H55834.MD
250 mg, Each
Quantity
Catalog number: H55834.MD
also known as H55834-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
402-46-0
IUPAC Name
4-fluorobenzene-1-sulfonamide
Molecular Formula
C6H6FNO2S
InChI Key
LFLSATHZMYYIAQ-UHFFFAOYSA-N
SMILES
NS(=O)(=O)C1=CC=C(F)C=C1
Assay (HPLC)
>98.0%

Description

4-Fluorobenzenesulfonamide, is used to produce bis-(4-fluoro-benzenesulfonyl)-amine with 4-fluoro-benzenesulfonyl chloride at temperature of 55 - 60. This reaction will need reagent OH-.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorobenzenesulfonamide, is used to produce bis-(4-fluoro-benzenesulfonyl)-amine with 4-fluoro-benzenesulfonyl chloride at temperature of 55 - 60. This reaction will need reagent OH-.

Solubility
It is soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text