Thermo Scientific Chemicals

2-(Tri-n-butylstannyl)oxazole, 95%, Thermo Scientific Chemicals

Catalog number: H55870.03
1 g, Each
Thermo Scientific Chemicals

2-(Tri-n-butylstannyl)oxazole, 95%, Thermo Scientific Chemicals

Catalog number: H55870.03
1 g, Each
Quantity
Catalog number: H55870.03
also known as H55870-03
Price (USD)
Have Questions?

Chemical Identifiers

CAS
145214-05-7
IUPAC Name
2-(tributylstannyl)-1,3-oxazole
Molecular Formula
C15H29NOSn
InChI Key
YOWGRWHKDCHINP-UHFFFAOYSA-N
SMILES
CCCC[Sn](CCCC)(CCCC)C1=NC=CO1
Refractive Index
1.493
Appearance (Color)
Colorless
Form
Liquid
Assay (GC)
≥94%
Infrared spectrum
Conforms to standard

Description

2-(Tri-n-butylstannyl)oxazole act as a synthetic building block, which is utilized in Stille coupling

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Tri-n-butylstannyl)oxazole act as a synthetic building block, which is utilized in Stille coupling

Solubility
Insoluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text