Thermo Scientific Chemicals

N-Isopropyl-4-methylbenzenesulfonamide, 97%, Thermo Scientific Chemicals

Catalog number: H55893.MB
25 mg, Each
Thermo Scientific Chemicals

N-Isopropyl-4-methylbenzenesulfonamide, 97%, Thermo Scientific Chemicals

Catalog number: H55893.MB
25 mg, Each
Quantity
Catalog number: H55893.MB
also known as H55893-MB
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
21230-07-9
IUPAC Name
4-methyl-N-(propan-2-yl)benzene-1-sulfonamide
Molecular Formula
C10H15NO2S
InChI Key
NXUHAAIJVABUMJ-UHFFFAOYSA-N
SMILES
CC(C)NS(=O)(=O)C1=CC=C(C)C=C1
Assay (HPLC)
>96.0%

Description

N-Isopropyl-4-methylbenzenesulfonamide, N-alkylation of sulfonamides using inactive ethers is used as alkylating reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Isopropyl-4-methylbenzenesulfonamide, N-alkylation of sulfonamides using inactive ethers is used as alkylating reagents.

Solubility
Soluble in water, ethanol.

Notes
Keep container tightly closed i
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text