It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Slightly soluble in water.
Notes
Store at room temperature. Avoid strong oxidizing agents.
RUO – Research Use Only
General References:
- James D. Kinder. Preparation and Structural Characterization of a Platinum Catecholate Complex Containing Two 3-Ethynylthiophene Groups. Organometallics. 1996, 15 (1),460-463.
- Jan Svoboda.; Jan Sedláček.; Jiří Zedník.; Gita Dvořáková.; Olga Trhlíková.; Dana Rédrová.; Hynek Balcar.; Jiří Vohlídal. Polymerization of 3-ethynylthiophene with homogeneous and heterogeneous Rh catalysts. J. Polym. Sci., Part A: Polym. Chem. 2008, 46 (8),2776-2787.