Thermo Scientific Chemicals

2-(Tri-n-butylstannyl)thiophene, 97%, Thermo Scientific Chemicals

Catalog number: H55904.AC
25 mL, Each
Thermo Scientific Chemicals

2-(Tri-n-butylstannyl)thiophene, 97%, Thermo Scientific Chemicals

Catalog number: H55904.AC
25 mL, Each
Quantity
Catalog number: H55904.AC
also known as H55904-AC
Price (USD)
Have Questions?

Chemical Identifiers

CAS
54663-78-4
Assay (GC)
97%

Description

2-(Tri-n-butylstannyl)thiophene derivative is employed in Stille reaction involving efficient carbon-carbon bond formation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Tri-n-butylstannyl)thiophene derivative is employed in Stille reaction involving efficient carbon-carbon bond formation.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text