3-Aminopropane-1-sulfonic acid, 97%, Thermo Scientific Chemicals
3-Aminopropane-1-sulfonic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Aminopropane-1-sulfonic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number H55918.06
also known as H55918-06
Price (USD)/ Each
116.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
116.00
Add to cart
3-Aminopropane-1-sulfonic acid, 97%, Thermo Scientific Chemicals
Catalog numberH55918.06
Price (USD)/ Each
116.00
-
Add to cart
Chemical Identifiers
CAS3687-18-1
IUPAC Name3-aminopropane-1-sulfonic acid
Molecular FormulaC3H9NO3S
InChI KeySNKZJIOFVMKAOJ-UHFFFAOYSA-N
SMILESNCCCS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Proton NMR97%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only