Thermo Scientific Chemicals

2-(1-Adamantyl)-4-methylphenol, 99%, Thermo Scientific Chemicals

Catalog number: H55922.03
1 g, Each
Thermo Scientific Chemicals

2-(1-Adamantyl)-4-methylphenol, 99%, Thermo Scientific Chemicals

Catalog number: H55922.03
1 g, Each
Quantity
Catalog number: H55922.03
also known as H55922-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
41031-50-9
IUPAC Name
2-(adamantan-1-yl)-4-methylphenol
Molecular Formula
C17H22O
InChI Key
XHLJIHBDAJFXBE-UHFFFAOYSA-N
SMILES
CC1=CC(=C(O)C=C1)C12CC3CC(CC(C3)C1)C2
Appearance (Color)
White
Form
Crystalline Powder

Description

2-(1-Adamantyl)-4-methylphenol, is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(1-Adamantyl)-4-methylphenol, is used as a pharmaceutical intermediate.

Solubility
Sparingly Soluble in water (7.8E-3 g/L) (25°C).

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text