2-(1-Adamantyl)-4-methylphenol, is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-(1-Adamantyl)-4-methylphenol, is used as a pharmaceutical intermediate.
Solubility
Sparingly Soluble in water (7.8E-3 g/L) (25°C).
Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only
General References:
- AG Dossetter.; TF Jamison.; EN Jacobsen. Highly Enantio-and Diastereoselective Hetero-Diels±Alder Reactions Catalyzed by New Chiral Tridentate Chromium (iii) Catalysts. catalyst.199938 (1), 163-165.
- P Kumar.;MS Bodas. A Novel Synthesis of 4 H-Chromen-4-ones via Intramolecular Wittig Reaction. Organic letters.200038 (1), 163-165.