4-Bromothiobenzamide, 97%, Thermo Scientific Chemicals
4-Bromothiobenzamide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Bromothiobenzamide, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H55956.03
also known as H55956-03
Price (USD)/ Each
19.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
22.90 
Save 3.25 (14%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
19.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
22.90 
Save 3.25 (14%)
Add to cart
4-Bromothiobenzamide, 97%, Thermo Scientific Chemicals
Catalog numberH55956.03
Price (USD)/ Each
19.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
22.90 
Save 3.25 (14%)
-
Add to cart
Specifications
Chemical Name or Material4-Bromothiobenzamide
CAS26197-93-3
Health Hazard 1H302-H317
Health Hazard 2GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.
Health Hazard 3P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only