Thermo Scientific Chemicals

1,1'-Di-n-octadecyl-3,3,3',3'-tetramethylindocarbocyanine perchlorate, 97%, Thermo Scientific Chemicals

Catalog number: H55957.MC
100 mg, Each
Thermo Scientific Chemicals

1,1'-Di-n-octadecyl-3,3,3',3'-tetramethylindocarbocyanine perchlorate, 97%, Thermo Scientific Chemicals

Catalog number: H55957.MC
100 mg, Each
Quantity
Catalog number: H55957.MC
also known as H55957-MC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
41085-99-8
IUPAC Name
2-[3-(3,3-dimethyl-1-octadecyl-2,3-dihydro-1H-indol-2-ylidene)prop-1-en-1-yl]-3,3-dimethyl-1-octadecyl-3H-indol-1-ium; perchloric acid
Molecular Formula
C59H98ClN2O4
InChI Key
JVXZRNYCRFIEGV-UHFFFAOYSA-N
SMILES
O[Cl](=O)(=O)=O.CCCCCCCCCCCCCCCCCCN1C2=CC=CC=C2C(C)(C)C1=CC=CC1=[N+](CCCCCCCCCCCCCCCCCC)C2=CC=CC=C2C1(C)C
Assay (HPLC)
>96.0%

Description

1,1'-Dioctadecyl-3,3,3',3'-tetramethylindocarbocyanine per chlorate is a lipophilic dye used to record membrane voltage for fluidity studies. This retrograde stain is useful in staining neurons because the stain is relatively nontoxic to cell bodies and the dye is retained in neurons. In addition, 1,1'-Dioctadecyl-3,3,3',3'-tetramethylindocarbocyanine per chlorate is an ideal retrograde and anterograde tracer to label the processes as well as the cell bodies of cultured neurons.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindocarbocyanine per chlorate is a lipophilic dye used to record membrane voltage for fluidity studies. This retrograde stain is useful in staining neurons because the stain is relatively nontoxic to cell bodies and the dye is retained in neurons. In addition, 1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindocarbocyanine per chlorate is an ideal retrograde and anterograde tracer to label the processes as well as the cell bodies of cultured neurons.

Solubility
Soluble in Dimethylformamide (DMF).

Notes
Reacts with strong oxidizing agents. Store at room temperature.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text