Thermo Scientific Chemicals

2-Acetyl-6-bromopyridine, 97%, Thermo Scientific Chemicals

Catalog number: H55981.06
5 g, Each
Thermo Scientific Chemicals

2-Acetyl-6-bromopyridine, 97%, Thermo Scientific Chemicals

Catalog number: H55981.06
5 g, Each
Quantity
Catalog number: H55981.06
also known as H55981-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
49669-13-8
IUPAC Name
1-(6-bromopyridin-2-yl)ethan-1-one
Molecular Formula
C7H6BrNO
InChI Key
RUJTWTUYVOEEFW-UHFFFAOYSA-N
SMILES
CC(=O)C1=NC(Br)=CC=C1
Assay (HPLC)
>96.0%

Description

2-Acetyl-6-bromopyridine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as OLED intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Acetyl-6-bromopyridine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as OLED intermediate.

Solubility
Soluble in methanol.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text