4-Chlorobenzeneboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
4-Chlorobenzeneboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Chlorobenzeneboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number H55984.06
also known as H55984-06
Price (USD)/ Each
88.30
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
88.30
Add to cart
4-Chlorobenzeneboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Catalog numberH55984.06
Price (USD)/ Each
88.30
-
Add to cart
Specifications
Chemical Name or Material4-Chlorobenzeneboronic acid pinacol ester
Melting Point50°C to 55°C
CAS195062-61-4
ColorBrown to White
Health Hazard 1H315-H319-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only