Thermo Scientific Chemicals

(R)-(+)-1-(2-Bromophenyl)ethanol, 98%, Thermo Scientific Chemicals

Catalog number: H56003.03
1 g, Each
Thermo Scientific Chemicals

(R)-(+)-1-(2-Bromophenyl)ethanol, 98%, Thermo Scientific Chemicals

Catalog number: H56003.03
1 g, Each
Quantity
Catalog number: H56003.03
also known as H56003-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
76116-20-6
IUPAC Name
(1R)-1-(2-bromophenyl)ethan-1-ol
Molecular Formula
C8H9BrO
InChI Key
DZLZSFZSPIUINR-ZCFIWIBFSA-N
SMILES
C[C@@H](O)C1=CC=CC=C1Br
Assay (unspecified)
>97.5%

Description

It is the most important dyestuff intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is the most important dyestuff intermediate.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text