Thermo Scientific Chemicals

Dicyclohexyl phthalate, 99+%, Thermo Scientific Chemicals

Catalog number: H56004.22
100 g, Each
Thermo Scientific Chemicals

Dicyclohexyl phthalate, 99+%, Thermo Scientific Chemicals

Catalog number: H56004.22
100 g, Each
Quantity
Catalog number: H56004.22
also known as H56004-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
84-61-7
IUPAC Name
1,2-dicyclohexyl benzene-1,2-dicarboxylate
Molecular Formula
C20H26O4
InChI Key
VOWAEIGWURALJQ-UHFFFAOYSA-N
SMILES
O=C(OC1CCCCC1)C1=CC=CC=C1C(=O)OC1CCCCC1
Assay (HPLC)
>99.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text