Dicyclohexyl phthalate, 99+%, Thermo Scientific Chemicals
Dicyclohexyl phthalate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dicyclohexyl phthalate, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number H56004.36
also known as H56004-36
Price (USD)/ Each
109.00
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
109.00
Add to cart
Dicyclohexyl phthalate, 99+%, Thermo Scientific Chemicals
Catalog numberH56004.36
Price (USD)/ Each
109.00
-
Add to cart
Chemical Identifiers
CAS84-61-7
IUPAC Name1,2-dicyclohexyl benzene-1,2-dicarboxylate
Molecular FormulaC20H26O4
InChI KeyVOWAEIGWURALJQ-UHFFFAOYSA-N
SMILESO=C(OC1CCCCC1)C1=CC=CC=C1C(=O)OC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>99.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only