Thermo Scientific Chemicals

Iodine trichloride, 97%, Thermo Scientific Chemicals

Catalog number: H56005.22
100 g, Each
Thermo Scientific Chemicals

Iodine trichloride, 97%, Thermo Scientific Chemicals

Catalog number: H56005.22
100 g, Each
Quantity
Catalog number: H56005.22
also known as H56005-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
865-44-1
IUPAC Name
trichloro-λ³-iodane
Molecular Formula
Cl3I
InChI Key
PAWIVBWALDNUJP-UHFFFAOYSA-N
SMILES
Cl[I](Cl)Cl
Assay (Redox Titration)
96.0% min

Description

Iodine trichloride can be used to introduce iodine and chlorine in organic synthesis. It is also used as topical antiseptic and as laboratory reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodine trichloride can be used to introduce iodine and chlorine in organic synthesis. It is also used as topical antiseptic and as laboratory reagent.

Solubility
Soluble with decomposition.

Notes
Light Sensitive. Store at -20°C.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text