1-(Benzyloxycarbonyl)piperazine, 98%, Thermo Scientific Chemicals
1-(Benzyloxycarbonyl)piperazine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-(Benzyloxycarbonyl)piperazine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
5 mL
25 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number H56008.AE
also known as H56008-AE
Price (USD)/ Each
816.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
960.00 
Save 143.35 (15%)
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
816.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
960.00 
Save 143.35 (15%)
Add to cart
1-(Benzyloxycarbonyl)piperazine, 98%, Thermo Scientific Chemicals
Catalog numberH56008.AE
Price (USD)/ Each
816.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
960.00 
Save 143.35 (15%)
-
Add to cart
Chemical Identifiers
CAS31166-44-6
IUPAC Name4-[(benzyloxy)carbonyl]piperazin-1-ium
Molecular FormulaC12H17N2O2
InChI KeyCTOUWUYDDUSBQE-UHFFFAOYSA-O
SMILESO=C(OCC1=CC=CC=C1)N1CC[NH2+]CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)97.5% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only