Thermo Scientific Chemicals

6-Bromooxindole, 97%, Thermo Scientific Chemicals

Catalog number: H56012.06
5 g, Each
Thermo Scientific Chemicals

6-Bromooxindole, 97%, Thermo Scientific Chemicals

Catalog number: H56012.06
5 g, Each
Quantity
Catalog number: H56012.06
also known as H56012-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
99365-40-9
IUPAC Name
6-bromo-2,3-dihydro-1H-indol-2-one
Molecular Formula
C8H6BrNO
InChI Key
JARRYVQFBQVOBE-UHFFFAOYSA-N
SMILES
BrC1=CC=C2CC(=O)NC2=C1
Assay (HPLC)
97%

Description

6-Bromooxindole derivative is used in the preparation of p38alpha inhibitors as potential anti- inflammatories. It is used as an intermediate in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Bromooxindole derivative is used in the preparation of p38alpha inhibitors as potential anti- inflammatories. It is used as an intermediate in pharmaceuticals.

Solubility
Slightly soluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text