Glycine benzyl ester p-toluenesulfonate salt is a potent crosslinking inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Glycine benzyl ester p-toluenesulfonate salt is a potent crosslinking inhibitor.
Solubility
Soluble in methanol. (0.1 g/mL)
Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
RUO – Research Use Only
Leonidas Zervas.; Milton Winitz.; Jesse P. Greenstein. Studies on Arginine Peptides. I. Intermediates in the Synthesis of N-Terminal and C-Terminal Arginine Peptides.J. Org. Chem. 1957, 22 (11),1515-1521 .
Robert L. Letsinger.; Milton J. Kornet. Popcorn Polymer as a Support in Multistep Syntheses.J. Am. Chem. Soc. 1963, 85 (19),3045-3046 .