Thermo Scientific Chemicals

Diphenyl-2-pyridylphosphine, 98%, Thermo Scientific Chemicals

Catalog number: H56029.14
25 g, Each
Thermo Scientific Chemicals

Diphenyl-2-pyridylphosphine, 98%, Thermo Scientific Chemicals

Catalog number: H56029.14
25 g, Each
Quantity
Catalog number: H56029.14
also known as H56029-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
37943-90-1
IUPAC Name
2-(diphenylphosphanyl)pyridine
Molecular Formula
C17H14NP
InChI Key
SVABQOITNJTVNJ-UHFFFAOYSA-N
SMILES
C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=N1
Appearance (Color)
Pale yellow
Assay from Suppliers CofA
≥97.5%
Form
Solid

Description

It is a ligand for metal-catalyzed carbonylations, hydration, dehydrogenative coupling, carbostannylation, distannylation, and silylation. It is also employed as a reagent for Mitsunobu reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a ligand for metal-catalyzed carbonylations, hydration, dehydrogenative coupling, carbostannylation, distannylation, and silylation. It is also employed as a reagent for Mitsunobu reaction.

Solubility
Soluble in toluene. (almost transparency)

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text