Thermo Scientific™

(R)-3-Isopropyl-2,5-dimethoxy-3,6-dihydropyrazine, 98%, Thermo Scientific Chemicals

Catalog number: H56077.XF
1 mL, Each
Thermo Scientific™

(R)-3-Isopropyl-2,5-dimethoxy-3,6-dihydropyrazine, 98%, Thermo Scientific Chemicals

Catalog number: H56077.XF
1 mL, Each
Quantity
Catalog number: H56077.XF
also known as H56077-XF
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
109838-85-9
IUPAC Name
(2R)-3,6-dimethoxy-2-(propan-2-yl)-2,5-dihydropyrazine
Molecular Formula
C9H16N2O2
InChI Key
FCFWEOGTZZPCTO-MRVPVSSYSA-N
SMILES
COC1=N[C@H](C(C)C)C(OC)=NC1
Appearance (Color)
Colorless to yellow to orang
Assay (GC)
≥97.5%
Proton NMR
Conforms
Form
Liquid

Description

(R)-3-Isopropyl-2,5-dimethoxy-3,6-dihydropyrazineChiral auxiliary for the synthesis of α-amino acids. It is an inhibitor with antitumor activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-3-Isopropyl-2,5-dimethoxy-3,6-dihydropyrazineChiral auxiliary for the synthesis of α-amino acids. It is an inhibitor with antitumor activity.

Solubility
Soluble in Chloroform and Ethanol

Notes
Store at -20°C. Keep away from oxidizing agents. Hygroscopic. Keep under inert atmosphere.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text