Thermo Scientific Chemicals

1,1,4,7,10,10-Hexamethyltriethylenetetramine, 97%, Thermo Scientific Chemicals

Catalog number: H56089.06
5 g, Each
Thermo Scientific Chemicals

1,1,4,7,10,10-Hexamethyltriethylenetetramine, 97%, Thermo Scientific Chemicals

Catalog number: H56089.06
5 g, Each
Quantity
Catalog number: H56089.06
also known as H56089-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3083-10-1
IUPAC Name
2,5,8,11-tetramethyl-2,5,8,11-tetraazadodecane
Molecular Formula
C12H30N4
InChI Key
DWFKOMDBEKIATP-UHFFFAOYSA-N
SMILES
CN(C)CCN(C)CCN(C)CCN(C)C
Assay (GC)
96.0% min

Description

1,1,4,7,10,10-Hexamethyltriethylenetetramine may be used as reagent in the synthesis of ideal linear random copolymers containing both vinyl polymer and polyester units in a single polymer chain.1 1,1,4,7,10,10-Hexamethyltriethylenetetramine complexed with CuBr constitutes catalytic complex, used in the copolymerization of poly[ε-caprolactone] with N,N-dimethylamino-2-ethyl methacrylate monomers by atom-transfer radical polymerization (ATRP).2 It may be used as catalyst in the aqueous surface-initiated-ATRP to grow poly(N,N-dimethylacrylamide) (PDMA).3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1,4,7,10,10-Hexamethyltriethylenetetramine may be used as reagent in the synthesis of ideal linear random copolymers containing both vinyl polymer and polyester units in a single polymer chain.1 1,1,4,7,10,10-Hexamethyltriethylenetetramine complexed with CuBr constitutes catalytic complex, used in the copolymerization of poly[ε-caprolactone] with N,N-dimethylamino-2-ethyl methacrylate monomers by atom-transfer radical polymerization (ATRP).2 It may be used as catalyst in the aqueous surface-initiated-ATRP to grow poly(N,N-dimethylacrylamide) (PDMA).3

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents. Store at room temperature.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text