Thermo Scientific Chemicals

2,7-Dibromonaphthalene, 99%, Thermo Scientific Chemicals

Catalog number: H56097.06
5 g, Each
Thermo Scientific Chemicals

2,7-Dibromonaphthalene, 99%, Thermo Scientific Chemicals

Catalog number: H56097.06
5 g, Each
Quantity
Catalog number: H56097.06
also known as H56097-06
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
58556-75-5
Assay (unspecified)
>98.5%

Description

It is an intermediate in dyestuffs and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an intermediate in dyestuffs and pharmaceuticals.

Solubility
Soluble in acetonitrile.

Notes
Incompatible with oxidizing agents. Store at room temperature.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Serious Eye Damage/Eye Irritation
    Category 2
    Chronic aquatic toxicity
    Category 2
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H319 - Causes serious eye irritation

    H411 - Toxic to aquatic life with long lasting effects

    Precautionary Statements

    P264 - Wash face, hands and any exposed skin thoroughly after handling

    P280 - Wear eye protection/ face protection

    P337 + P313 - If eye irritation persists: Get medical advice/attention