Thermo Scientific Chemicals

2,4-Dichloro-5-methoxypyrimidine, 97%, Thermo Scientific Chemicals

Catalog number: H56153.09
10 g, Each
Thermo Scientific Chemicals

2,4-Dichloro-5-methoxypyrimidine, 97%, Thermo Scientific Chemicals

Catalog number: H56153.09
10 g, Each
Quantity
Catalog number: H56153.09
also known as H56153-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
19646-07-2
Appearance (Color)
White
Proton NMR
Conforms to structure
Form
Crystalline powder
Assay (GC)
>96.0%

Description

It is used for preparation of heteroarylpiperazine derivatives for use in treatment of Alzheimer's disease.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used for preparation of heteroarylpiperazine derivatives for use in treatment of Alzheimer’s disease.

Solubility
Slightly soluble in water.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text