Thermo Scientific Chemicals

5-Chlorovaleroyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: H56237.18
50 g, Each
Thermo Scientific Chemicals

5-Chlorovaleroyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: H56237.18
50 g, Each
Quantity
Catalog number: H56237.18
also known as H56237-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1575-61-7
IUPAC Name
5-chloropentanoyl chloride
Molecular Formula
C5H8Cl2O
InChI Key
SVNNWKWHLOJLOK-UHFFFAOYSA-N
SMILES
ClCCCCC(Cl)=O
Assay (unspecified)
>97.5%

Description

5-Chlorovaleroyl chloride (5-CVC) has been used in the development and validation of a GC-FID method for the analysis of low level impurities present in 5-CVC. It has been used in the synthesis of anion-exchange resins by acylation of polystyrene/divinylbenzene and subsequent amination with trimethylamine. It is commonly used as alkylating agent in the synthesis of pharmaceutical intermediates and other specialty chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chlorovaleroyl chloride (5-CVC) has been used in the development and validation of a GC-FID method for the analysis of low level impurities present in 5-CVC. It has been used in the synthesis of anion-exchange resins by acylation of polystyrene/divinylbenzene and subsequent amination with trimethylamine. It is commonly used as alkylating agent in the synthesis of pharmaceutical intermediates and other specialty chemicals.

Solubility
Reacts with water.

Notes
Moisture sensitive. Incompatible with oxidizing agents, bases, alcohols, amines.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text