Thermo Scientific Chemicals

Pentacene, 97%, Thermo Scientific Chemicals

Catalog number: H56310.MD
250 mg, Each
Thermo Scientific Chemicals

Pentacene, 97%, Thermo Scientific Chemicals

Catalog number: H56310.MD
250 mg, Each
Quantity
Catalog number: H56310.MD
also known as H56310-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
135-48-8
IUPAC Name
pentacene
Molecular Formula
C22H14
InChI Key
SLIUAWYAILUBJU-UHFFFAOYSA-N
SMILES
C1=CC2=CC3=CC4=CC5=CC=CC=C5C=C4C=C3C=C2C=C1
Appearance (Color)
Dark blue to black
Form
Solid
Assay (unspecified)
>96.0%

Description

Pentacene can be used as OPV device (Organo Photovoltaic) and as semiconductors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentacene can be used as OPV device (Organo Photovoltaic) and as semiconductors.

Solubility
Soluble in organic solvents.

Notes
Air and light sensitive. Store at cool temperature.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text