Thermo Scientific Chemicals

9,9-Di-n-dodecyl-2,7-dibromofluorene, 98%, Thermo Scientific Chemicals

Catalog number: H56320.22
100 g, Each
Thermo Scientific Chemicals

9,9-Di-n-dodecyl-2,7-dibromofluorene, 98%, Thermo Scientific Chemicals

Catalog number: H56320.22
100 g, Each
Quantity
Catalog number: H56320.22
also known as H56320-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
286438-45-7
IUPAC Name
2,7-dibromo-9,9-didodecyl-9H-fluorene
Molecular Formula
C37H56Br2
InChI Key
KFOUJVGPGBSIFB-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCC1(CCCCCCCCCCCC)C2=C(C=CC(Br)=C2)C2=C1C=C(Br)C=C2
Assay (HPLC)
>97.5%

Description

It is an intermediate for polymeric light-emitting diodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an intermediate for polymeric light-emitting diodes.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text