Thermo Scientific Chemicals

5,5'-Dibromo-2,2'-bithiophene, 98%, Thermo Scientific Chemicals

Catalog number: H56337.14
25 g, Each
Thermo Scientific Chemicals

5,5'-Dibromo-2,2'-bithiophene, 98%, Thermo Scientific Chemicals

Catalog number: H56337.14
25 g, Each
Quantity
Catalog number: H56337.14
also known as H56337-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4805-22-5
IUPAC Name
5,5'-dibromo-2,2'-bithiophene
Molecular Formula
C8H4Br2S2
InChI Key
SXNCMLQAQIGJDO-UHFFFAOYSA-N
SMILES
BrC1=CC=C(S1)C1=CC=C(Br)S1
Appearance (Color)
White
Form
Powder or crystals or crystalline powder
Assay from Supplier's CofA
≥97.5%

Description

It is an active pharmaceutical ingredient and an OLED (organic light-emitting diode) intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical ingredient and an OLED (organic light-emitting diode) intermediate.

Solubility
Slightly soluble in water.

Notes
Recommended storage temperature: 2 - 4°C.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text