Thermo Scientific Chemicals

(R)-(-)-O-Acetylmandelic acid, 98%, Thermo Scientific Chemicals

Catalog number: H56492.03
1 g, Each
Thermo Scientific Chemicals

(R)-(-)-O-Acetylmandelic acid, 98%, Thermo Scientific Chemicals

Catalog number: H56492.03
1 g, Each
Quantity
Catalog number: H56492.03
also known as H56492-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
51019-43-3
IUPAC Name
2-(acetyloxy)-2-phenylacetic acid
Molecular Formula
C10H10O4
InChI Key
OBCUSTCTKLTMBX-UHFFFAOYNA-N
SMILES
CC(=O)OC(C(O)=O)C1=CC=CC=C1
Assay (HPLC)
>97.5%

Description

The (R)- and (S)-isomers are chiral derivatizing agents for NMR determination of enantiomeric purity of α-deuterated carboxylic acids, alcohols, and amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The (R)- and (S)-isomers are chiral derivatizing agents for NMR determination of enantiomeric purity of α-deuterated carboxylic acids, alcohols, and amines.

Solubility
Soluble in methanol. (almost transparency)

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text