N-Boc-1,4-diaminobutane is used in preparation of pharmacologically active compounds, preparation of spermidine analogues and in introduction of a C4-spacer. It serve as intermediates of medicine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Boc-1,4-diaminobutane is used in preparation of pharmacologically active compounds, preparation of spermidine analogues and in introduction of a C4-spacer. It serve as intermediates of medicine.
Notes
Incompatible with oxidizing agents. Store at room temperature.
RUO – Research Use Only
R. Jin.; L.S. Moreira Teixeira.; P.J. Dijkstra.; C.A. van Blitterswijk.; M. Karperien.; J. Feijena. Enzymatically-crosslinked injectable hydrogels based on biomimetic dextran-hyaluronic acid conjugates for cartilage tissue engineering.Biomaterials. 2010, 31 (11), 3103-3113.
Jerry Ryan Holder.; Rayna M Bauzo.; Zhimin Xiang, Joseph Scott.; Carrie Haskell-Luevano. Design and pharmacology of peptoids and peptide-peptoid hybrids based on the melanocortin agonists core tetrapeptide sequence. Bioorg. Med. Chem. Lett. 2003, 13 (24),4505-4509.