Thermo Scientific Chemicals

n-Butyl nitrite, 95%, Thermo Scientific Chemicals

Catalog number: H56501.36
500 g, Each
Thermo Scientific Chemicals

n-Butyl nitrite, 95%, Thermo Scientific Chemicals

Catalog number: H56501.36
500 g, Each
Quantity
Catalog number: H56501.36
also known as H56501-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
544-16-1
IUPAC Name
butyl nitrite
Molecular Formula
C4H9NO2
InChI Key
JQJPBYFTQAANLE-UHFFFAOYSA-N
SMILES
CCCCON=O
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay from Suppliers CofA
≥94.0%

Description

It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text