Diethyl cyclopropane-1,1-dicarboxylate, Thermo Scientific Chemicals
Diethyl cyclopropane-1,1-dicarboxylate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethyl cyclopropane-1,1-dicarboxylate, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H56568.03
also known as H56568-03
Price (USD)/ Each
32.60
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
32.60
Add to cart
Diethyl cyclopropane-1,1-dicarboxylate, Thermo Scientific Chemicals
Catalog numberH56568.03
Price (USD)/ Each
32.60
-
Add to cart
Chemical Identifiers
CAS1559-02-0
IUPAC Name1,1-diethyl cyclopropane-1,1-dicarboxylate
Molecular FormulaC9H14O4
InChI KeyKYYUCZOHNYSLFV-UHFFFAOYSA-N
SMILESCCOC(=O)C1(CC1)C(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay from Supplier's CofA≥95.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only