Thermo Scientific Chemicals

2,3-Dimercapto-1-propanesulfonic acid sodium salt monohydrate, 95%, Thermo Scientific Chemicals

Catalog number: H56578.03
1 g, Each
Thermo Scientific Chemicals

2,3-Dimercapto-1-propanesulfonic acid sodium salt monohydrate, 95%, Thermo Scientific Chemicals

Catalog number: H56578.03
1 g, Each
Quantity
Catalog number: H56578.03
also known as H56578-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
207233-91-8
IUPAC Name
(2R)-2,3-disulfanylpropane-1-sulfonate
Molecular Formula
C3H7O3S3
InChI Key
JLVSRWOIZZXQAD-GSVOUGTGSA-M
SMILES
[O-]S(=O)(=O)C[C@H](S)CS
Assay (unspecified)
≥94.0%

Description

Sodium 2,3-dimercaptopropanesulfonate monohydrate has been used in a study to assess its use as an antidote.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium 2,3-dimercaptopropanesulfonate monohydrate has been used in a study to assess its use as an antidote.

Solubility
Soluble in Water: 100 mg/ml

Notes
Store at 4°C. Incompatible with oxidizing agents. Keep container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text