Thermo Scientific Chemicals

(2-Bromoethoxy)-tert-butyldimethylsilane, 98%, stab. with sodium carbonate, Thermo Scientific Chemicals

Catalog number: H56594.06
5 g, Each
Thermo Scientific Chemicals

(2-Bromoethoxy)-tert-butyldimethylsilane, 98%, stab. with sodium carbonate, Thermo Scientific Chemicals

Catalog number: H56594.06
5 g, Each
Quantity
Catalog number: H56594.06
also known as H56594-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
86864-60-0
IUPAC Name
(2-bromoethoxy)(tert-butyl)dimethylsilane
Molecular Formula
C8H19BrOSi
InChI Key
JBKINHFZTVLNEM-UHFFFAOYSA-N
SMILES
CC(C)(C)[Si](C)(C)OCCBr
Appearance (Color)
Clear
Form
Liquid
Assay (HPLC)
≥97.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text