Thermo Scientific Chemicals

Cyclopentanecarboxaldehyde, 98%, stab., Thermo Scientific Chemicals

Catalog number: H56609.06
5 g, Each
Thermo Scientific Chemicals

Cyclopentanecarboxaldehyde, 98%, stab., Thermo Scientific Chemicals

Catalog number: H56609.06
5 g, Each
Quantity
Catalog number: H56609.06
also known as H56609-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
872-53-7
IUPAC Name
cyclopentanecarbaldehyde
Molecular Formula
C6H10O
InChI Key
VELDYOPRLMJFIK-UHFFFAOYSA-N
SMILES
O=CC1CCCC1
Appearance (Color)
Clear colorless
Elemental Analysis
>97.5%
Proton NMR
Conforms
Stabilizer
w/0.1% hydroquinone or with 25 ppm 1,4-benzoquinone
Form
Liquid

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Soluble in water and ethanol

Notes
Light sensitive. Incompatible with oxidizing agents. Store at room temperature.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text