Thermo Scientific Chemicals

3,4-Dimethoxythiophene, 98%, Thermo Scientific Chemicals

Catalog number: H56674.03
1 g, Each
Thermo Scientific Chemicals

3,4-Dimethoxythiophene, 98%, Thermo Scientific Chemicals

Catalog number: H56674.03
1 g, Each
Quantity
Catalog number: H56674.03
also known as H56674-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
51792-34-8
IUPAC Name
3,4-dimethoxythiophene
Molecular Formula
C6H8O2S
InChI Key
ZUDCKLVMBAXBIF-UHFFFAOYSA-N
SMILES
COC1=CSC=C1OC
Assay (GC)
98%

Description

3,4-Dimethoxythiophene is used as an electronic materials intermediate. It is used as a starting material in the synthesis of porphyrin dyad which is used to study photoinduced energy transfer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethoxythiophene is used as an electronic materials intermediate. It is used as a starting material in the synthesis of porphyrin dyad which is used to study photoinduced energy transfer.

Solubility
Miscible with organic solvents. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text