4-Chloroquinoline, 99%, Thermo Scientific Chemicals
4-Chloroquinoline, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Chloroquinoline, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H56721.06
also known as H56721-06
Price (USD)/ Each
372.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
372.00
Add to cart
4-Chloroquinoline, 99%, Thermo Scientific Chemicals
Catalog numberH56721.06
Price (USD)/ Each
372.00
-
Add to cart
Chemical Identifiers
CAS611-35-8
IUPAC Name4-chloroquinoline
Molecular FormulaC9H6ClN
InChI KeyKNDOFJFSHZCKGT-UHFFFAOYSA-N
SMILESClC1=C2C=CC=CC2=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>98.5%
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Partly soluble in water.

Notes
Incompatible with oxidizing agents. Store at room temperature.
RUO – Research Use Only

General References:

  1. O. G. Backeberg. The reaction between phenylhydrazine and 4-chloroquinoline derivatives, and the preparation of the corresponding 4-benzeneazo- and 4-amino-compounds.J. Chem. Soc. 1938, (0),1083-1087.
  2. Erwan Le Gall.; Corinne Gosmini.; Jean-Yves Nédélec.; Jacques Périchon. Cobalt-catalyzed electrochemical cross-coupling of functionalized phenyl halides with 4-chloroquinoline derivatives. Tetrahedron Lett. 2001 42 (2),267-269.