Thermo Scientific Chemicals

Bromomethyl acetate, 95%, Thermo Scientific Chemicals

Catalog number: H56755.09
10 g, Each
Thermo Scientific Chemicals

Bromomethyl acetate, 95%, Thermo Scientific Chemicals

Catalog number: H56755.09
10 g, Each
Quantity
Catalog number: H56755.09
also known as H56755-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
590-97-6
IUPAC Name
bromomethyl acetate
Molecular Formula
C3H5BrO2
InChI Key
NHYXMAKLBXBVEO-UHFFFAOYSA-N
SMILES
CC(=O)OCBr
Assay (GC)
95%

Description

Bromomethyl acetate is involved in the preparation of optically active cyclohexene antisepsis agents. It acts as reagent in samarium diiodide-mediated conversion of ketones and aldehydes to 1,2-diacetates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromomethyl acetate is involved in the preparation of optically active cyclohexene antisepsis agents. It acts as reagent in samarium diiodide-mediated conversion of ketones and aldehydes to 1,2-diacetates.

Solubility
Miscible with chloroform.

Notes
Store in cool place. Moisture sensitive and hygroscopic. Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text