Thermo Scientific Chemicals

4-Nitrophenyl beta-D-galactopyranoside, 98+%, Thermo Scientific Chemicals

Catalog number: H56905.06
5 g, Each
Thermo Scientific Chemicals

4-Nitrophenyl beta-D-galactopyranoside, 98+%, Thermo Scientific Chemicals

Catalog number: H56905.06
5 g, Each
Quantity
Catalog number: H56905.06
also known as H56905-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3150-24-1
IUPAC Name
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol
Molecular Formula
C12H15NO8
InChI Key
IFBHRQDFSNCLOZ-YBXAARCKSA-N
SMILES
OC[C@H]1O[C@@H](OC2=CC=C(C=C2)[N+]([O-])=O)[C@H](O)[C@@H](O)[C@H]1O
Assay (HPLC)
>98.0%

Description

It is useful in detecting the glycosylated enzyme.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is useful in detecting the glycosylated enzyme.

Solubility
Soluble in  DMSO, Methanol, Water.

Notes
Store at -20°C. Incompatible with oxidizing agents and heat.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text