It is useful in detecting the glycosylated enzyme.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is useful in detecting the glycosylated enzyme.
Solubility
Soluble in DMSO, Methanol, Water.
Notes
Store at -20°C. Incompatible with oxidizing agents and heat.
RUO – Research Use Only
R. Prasanna.; S. Pattnaik.; T. C. K Sugitha, L. Nain.; A. K. Saxena. Development of cyanobacterium-based biofilms and their in vitro evaluation for agriculturally useful traits. Folia Microbiologica. 2011, 56 (1),.49-58
Mangala Srinivas.; Philipp Boehm-Sturm.; Carl G. Figdor.; I. Jolanda de Vries.; Mathias Hoehn. Labeling cells for in vivo tracking using 19F MRI. Biomaterials. 2012, 33 (34),8830-8840.