It is used in the synthesis of novel glycyrrhetinic acid derivatives on il-1β-induced prostaglandin e2production in normal human dermal fibroblasts and also in the total synthesis of 6,7—Dihydroligustilide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in the synthesis of novel glycyrrhetinic acid derivatives on il-1β-induced prostaglandin e2production in normal human dermal fibroblasts and also in the total synthesis of 6,7—Dihydroligustilide.
Solubility
Reacts with water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
- Xue-Chuan Lv, et al. Molar heat capacity and thermodynamic properties of 1-cyclohexene-1,2-dicarboxylic anhydride [C8H8O3].J. Chem. Thermodyn.,2004,36(9), 787-792.
- T. S. Nimry, et al. Photoadducts of 1-cyclohexene-1,2-dicarboxylic anhydride.J. Org. Chem.,1983,48(22), 4102-4104.