4-Bromo-1-indanone, 97%, Thermo Scientific Chemicals
4-Bromo-1-indanone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Bromo-1-indanone, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
Catalog number H56937.06
also known as H56937-06
Price (USD)/ Each
137.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
137.00
Add to cart
4-Bromo-1-indanone, 97%, Thermo Scientific Chemicals
Catalog numberH56937.06
Price (USD)/ Each
137.00
-
Add to cart
Chemical Identifiers
CAS15115-60-3
IUPAC Name4-bromo-2,3-dihydro-1H-inden-1-one
Molecular FormulaC9H7BrO
InChI KeyUVVYFYLSZIMKMC-UHFFFAOYSA-N
SMILESBrC1=CC=CC2=C1CCC2=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only