9,9-Di-n-hexyl-2,7-dibromofluorene, 98%, Thermo Scientific Chemicals
9,9-Di-n-hexyl-2,7-dibromofluorene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

9,9-Di-n-hexyl-2,7-dibromofluorene, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number H56952.14
also known as H56952-14
Price (USD)/ Each
203.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
203.00
Add to cart
9,9-Di-n-hexyl-2,7-dibromofluorene, 98%, Thermo Scientific Chemicals
Catalog numberH56952.14
Price (USD)/ Each
203.00
-
Add to cart
Chemical Identifiers
CAS189367-54-2
IUPAC Name2,7-dibromo-9,9-dihexyl-9H-fluorene
Molecular FormulaC25H32Br2
InChI KeyOXFFIMLCSVJMHA-UHFFFAOYSA-N
SMILESCCCCCCC1(CCCCCC)C2=C(C=CC(Br)=C2)C2=C1C=C(Br)C=C2
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
It finds its application as an intermediate for polymeric light-emitting diodes and as an OLED material because of their pure blue emission and efficient electroluminescence coupled with a high charge-carrier mobility and good process ability.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application as an intermediate for polymeric light-emitting diodes and as an OLED material because of their pure blue emission and efficient electroluminescence coupled with a high charge-carrier mobility and good process ability.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. V. N. Bliznyuk and S. A. Carter. Electrical and Photo induced Degradation of Polyfluorene Based Films and Light-Emitting Devices.Macromolecules.,1999,32(2), 361-369.
  2. C. M. Martin, et al. Hydrostatic pressure dependence of the luminescence and Raman frequencies in polyfluorene.Phys. Rev. B.,2003,68(11), 115203.