It is applied in High Performance Liquid Chromatography. (HPLC) and Nuclear Magnetic Resonance (NMR).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is applied in High Performance Liquid Chromatography. (HPLC) and Nuclear Magnetic Resonance (NMR).
Solubility
Slightly soluble in water
Notes
Store at 2-8°C. Incompatible with oxidizing agents and heat.
RUO – Research Use Only
General References:
- Stefanie N. Guntari.; Aaron C. H. Khin.; Edgar H. H. Wong.; Tor K. Goh.; Anton Blencowe.; Frank Caruso.; Greg G. Qiao. (Super)hydrophobic and Multilayered Amphiphilic Films Prepared by Continuous Assembly of Polymers.Adv. Funct. Mater. 2013, 23 (41),5159-5166.
- Stefanie N. Guntari.; Tor K. Goh.; Anton Blencowe.; Edgar H. H. Wong.; Frank Caruso.; Greg G. Qiao. Factors influencing the growth and topography of nanoscale films fabricated by ROMP-mediated continuous assembly of polymers. Polym. Chem. 2013, 4 (1),68-75.