Thermo Scientific Chemicals

5-Norbornen-2-ol, mixture of endo and exo, 98+%, Thermo Scientific Chemicals

Catalog number: H56994.14
25 g, Each
Thermo Scientific Chemicals

5-Norbornen-2-ol, mixture of endo and exo, 98+%, Thermo Scientific Chemicals

Catalog number: H56994.14
25 g, Each
Quantity
Catalog number: H56994.14
also known as H56994-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13080-90-5
IUPAC Name
bicyclo[2.2.1]hept-5-en-2-ol
Molecular Formula
C7H10O
InChI Key
MKOSBHNWXFSHSW-UHFFFAOYNA-N
SMILES
OC1CC2CC1C=C2
Appearance (Color)
Pale cream to pale brown
Assay (GC)
≥ 98%
Form
Powder

Description

It is applied in High Performance Liquid Chromatography. (HPLC) and Nuclear Magnetic Resonance (NMR).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied in High Performance Liquid Chromatography. (HPLC) and Nuclear Magnetic Resonance (NMR).

Solubility
Slightly soluble in water

Notes
Store at 2-8°C. Incompatible with oxidizing agents and heat.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text