Thermo Scientific™

Ethyl (R)-(+)-1-Boc-4-oxopiperidine-2-carboxylate, 95%, Thermo Scientific™

Catalog number: H57019.03
1 g, Each
Thermo Scientific™

Ethyl (R)-(+)-1-Boc-4-oxopiperidine-2-carboxylate, 95%, Thermo Scientific™

Catalog number: H57019.03
1 g, Each
Quantity
Catalog number: H57019.03
also known as H57019-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
357154-16-6
IUPAC Name
1-tert-butyl 2-ethyl 4-oxopiperidine-1,2-dicarboxylate
Molecular Formula
C13H21NO5
InChI Key
YAVQLRUBKDCOCI-UHFFFAOYNA-N
SMILES
CCOC(=O)C1CC(=O)CCN1C(=O)OC(C)(C)C
Appearance (Color)
pale cream / Pale yellow
Proton NMR
confirms
Form
Low melting solid
Assay (GC)
?94%

Description

It is used as pharmaceutical intermediate.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as pharmaceutical intermediate.

Solubility
Soluble in chloroform and methanol.

Notes
Store in a cool ventilated place. Avoid strong oxidizing materials.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text